missing translation for 'onlineSavingsMsg'
Learn More

Melford Ficoll

Product Code. 30018265
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018265 5 g 5g
30018264 25 g 25g
30018263 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018265 Supplier Melford Supplier No. F104005.0
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ficoll is an uncharged polymer of sucrose widely used for stabilizing cells and isolation of nuclei and organelles.

Ficoll is an uncharged polymer of sucrose widely used for stabilizing cells and isolation of nuclei and organelles. Also used in Denhardt's Reagent and in gel loading buffers.

Specifications

Chemical Name or Material Ficoll
CAS 26873-85-8
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.