Learn More
Description
The NanoPhotometer™ field kit is your mobile screening lab. With up to 8 hours of battery power there is no need to worry about a power outlet. The convenient NanoPhotometer™ roller case is specially designed to fit into the overhead bins on most major airlines. To comply with TSA requirements the case can be equipped with a TSA lock for safe airline transportation.
- Outside Dimensions 480 x 380 x 240 mm
- Inside Dimensions 460 x 360 x 210 mm
- Weight 5.5 kg
- Total height when handle fully extended 1040 mm
- Inserts Custom fitted cross-linked foam, type: PLZ LD 45
Specifications
Specifications
| Description | Outside Dimensions - 480 x 380 x 240 mm, Inside Dimensions - 460 x 360 x 210 mm |
| Type | Field Kit for the NanoPhotometer |
| For Use With (Application) | Spectroscopy Anywhere (Up to 8 hours of battery power) |
| Holds | Pipette, Sample and Buffer Containers, Accessories and Documentation |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.