missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Filling Valve

Product Code. 15633977 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
25, 50, 100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15633977 25, 50, 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15633977 Supplier Brand™ Supplier No. 6698
  • ONLINE EXCLUSIVE PRICE
    25.60 €
    23.03 € / Pack of 1
    Save 2.57 €
    10% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Filling Valve
Volume (Metric) 25, 50, 100 mL
For Use With (Equipment) Dispensette III
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.