missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Replacement filling valves for BRAND™ Bottletop Dispensers

Product Code. 15693977 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
Dispensette S or S Organic - 1, 2, 5 and 10 mL
Dispensette S/S Organic - 25, 50, and 100 mL
Product Type:
Filling Valve
Includes:
Filling Valve
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Product Type Includes Unit Size
15693977 Dispensette S or S Organic - 1, 2, 5 and 10 mL Filling Valve Filling Valve Each
15533583 Dispensette S/S Organic - 25, 50, and 100 mL Filling Valve Filling Valve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15693977 Supplier BRAND™ Supplier No. 6734
  • ONLINE EXCLUSIVE PRICE
    21.80 €
    19.59 € / Pack of 1
    Save 2.21 €
    10% Off
View all promotions
In Stock
Add to basket
Lägg i varukorgen
Bulk Quote Request

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Filling valves and tools available for the proper cleaning and maintenance of various BRAND™ bottletop dispensers.

  • Filling valves for the Dispensette™ III, Organic, TA and seripettor™ pro are provided with a sealing washer. The updated design of the Dispensette™ S series of bottletop dispensers has eliminated the need for a sealing washer.
  • Filling valve with olive-shaped nozzle is recommended for Dispensette™ III and Organic bottletop instruments that are frequently autoclaved.
  • seripettor™ valve set includes both the filling and discharge valves for the instrument.
  • Calibrating-mounting tool is used for calibrating analog-adjustable Dispensette™ models, as well as removing the valves for routine cleaning or replacement.
TRUSTED_SUSTAINABILITY

Specifikationer

Product Type Filling Valve
Autoclavable Autoclavable
Color Translucent/White
Material ETFE, Borosilicate Glass, Ceramic
For Use With (Equipment) Dispensette S or S Organic - 1, 2, 5 and 10 mL
Includes Filling Valve
Sterility Non-sterile
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.