Learn More
CELLPATH FILOSLIDE 10 SLIDE TRAY
Product Code. 30226317
Change view
Click to view available options
Color:
Blue
Red
White
Yellow
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30226317 White Pack of 10
30226318 Blue Pack of 10
30226319 Yellow Pack of 10
30226320 Red Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30226317 Supplier CELLPATH Supplier No. WEX110201B
  • ONLINE EXCLUSIVE PRICE
    101.88 €
    42.64 € / Pack of 10
    Save 59.24 €
    58% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The FiloSlide 10 slide trays feature a useful 360° cover flap, which neatly folds back on itself to save working space. An index reference is included on the flap and a recessed well on the inside of this flap prevents damp slides from sticking to the board.
  • The FiloSlide 10 tray, which measures 335 mm x 102 mm has a single column of 10 wells.
  • Each slide has its own easy release compartment which is individually numbered.
  • Constructed from solid board and available in six colours including black, white, yellow, blue, green and red.
  • This allows laboratories to colour code different types of workload or different applications.
  • Key Benefits:
  • 360° cover flap saves working space requirement
  • Range of colours available to assist in team resource allocation
  • Thumb grooves for easy removal of slides
  • Robust, allowing multiple use and cost savings
Specifications
Type Slide Holder
Slide Capacity 10
Color White