Learn More
CELLPATH FILOSLIDE 100 BOX
Product Code. 30226306
Change view
Click to view available options
Color:
Black
Blue
Orange
White
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30226306 White Each
30226305 Black Each
30226307 Blue Each
30226308 Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30226306 Supplier CELLPATH Supplier No. WEQ040200A
  • ONLINE EXCLUSIVE PRICE
    33.09 €
    13.84 € / Pack of 1
    Save 19.25 €
    58% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
This sturdy FiloSlide Box offers safe and cost-effective storage for microscope slides.
  • Key Benefits:
  • Stackable
  • Snap closure to seal out dust
  • Splash-proof
  • Double hinged lid prevents loss or breakage
  • Index card included for easy specimen identification
  • Can store up to 100 micrscope slides
Specifications
Type Slide Box
Slide Capacity 100
Color White