Learn More
CELLPATH FILOSLIDE 25 BOX
Product Code. 30226299
Change view
Click to view available options
Color:
Black
Blue
Orange
Red
White
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30226299 Blue Each
30226297 Black Each
30226298 White Each
30226300 Orange Each
30226301 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30226299 Supplier CELLPATH Supplier No. WEQ020600A
  • ONLINE EXCLUSIVE PRICE
    19.23 €
    8.05 € / Pack of 1
    Save 11.18 €
    58% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This sturdy FiloSlide Box offers safe and cost-effective storage for microscope slides.
  • Key Benefits:
  • Stackable
  • Snap closure to seal out dust
  • Splash-proof
  • Double hinged lid prevents loss or breakage
  • Index card included for easy specimen identification
  • Can store up to 25 micrscope slides
Specifications
Type Slide Box
Slide Capacity 25
Color Blue