missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Filter Boards, Discs, Grade C 160

Filter boards with increased wet strength for the filtration and absorption of liquids.

Brand:  Sartorius Stedim Biotech™ FT-3-343-150

Additional Details : Weight : 0.14100kg

 View more versions of this product

Product Code. 15932930

  • 18.80 € / Pack of 50
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

  • Constructed of cotton linters and refined pulp
  • Suitable for analytical sample preparation
  • Non-sterile, bulk packed
Specifications

Specifications

160 g/m2
Cotton Linters, Refined Pulp
50/Pk.
150 mm
Disc
C 160
0.3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers