missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fimepinostat, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Description
Fimepinostat (CUDC-907) potently inhibits class I PI3Ks as well as classes I and II HDAC enzymes with an IC50 of 19/54/39 nM and 1.7/5.0/1.8/2.8 nM for PI3Kα/PI3Kβ/PI3Kδ and HDAC1/HDAC2/HDAC3/HDAC10 , respectively.
Specifications
Specifications
| Chemical Name or Material | Fimepinostat |
| CAS | 1339928-25-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H24N8O4S |
| Quantity | 50 mg |
| Synonym | CUDC-907 |
| Solubility Information | DMSO : 50 mg/mL (98.32 mM; Need ultrasonic) ∣DMF : 5 mg/mL (9.83 mM; Need ultrasonic) |
| SMILES | O=C(C1=CN=C(N(CC2=CC3=NC(C4=CC=C(OC)N=C4)=NC(N5CCOCC5)=C3S2)C)N=C1)NO |
| Molecular Weight (g/mol) | 508.55 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction