missing translation for 'onlineSavingsMsg'
Learn More

Heinz Herenz Fine Tip LMS-Swab Thio Broth

Product Code. 30158308
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 150
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
30158308 Sterile Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30158308 Supplier Heinz Herenz Supplier No. 1020313
  • ONLINE EXCLUSIVE PRICE
    705.60 €
    456.39 € / Pack of 150
    Save 249.21 €
    35% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains 2 mL of liquid thio broth medium. Sterile.

Our transport and collection swabs contain special applicators and test tubes with different liquid media, customized for the transportation and preservation of microbiological samples.

  • The liquid medium allows the division of the sample material and thus several tests from one sample.
  • The ready to use devices are supplied in a peelpack which is easy to open.
  • The breakpoint in the shaft of the applicator enables a safe and hygienic workflow.
  • The standard size of the test tubes with screwcap simplified the use with automatic analyzers.
  • There are labels on the test tubes for a accurate sample identification.
  • The items are produced according to the conditions of GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485. This ensures a high quality.
  • Our swabs are sterile (SAL 106), pyrogen free and noncytotoxic.

Specifications

Product Type Transport and Collection Swab
Format Swab
Sterility Sterile
Color White
compliance-icons
Product Identifier
  • X150 LMS-Swab Thio Broth, foam swab, standard tip, black screw cap, 2ml of Thio Broth medium
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: sodium thioglycolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.