missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Finedex™ Latex Gloves

Product Code. 12645213
Change view
Click to view available options
Size:
9
10
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12645213 9 Each
12605213 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12645213 Supplier Honeywell Safety Products™ Supplier No. 209440109
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Flexible latex, comfortable cotton flock, and a diamond-pattern finish make this glove a useful and practical choice for a variety of needs. Honeywell Safety Products™ Finedex™ Latex Gloves are manufactured via a process that minimizes latex protein.

  • Natural latex glove for flexibility, dexterity, and good mechanical and chemical resistance against detergents.
  • Cotton flock interior for comfort.
  • Diamond-pattern finish for grasping dry and damp objects.
  • Cuff protects forearm.
  • Minimum latex protein.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 374-2003 Chemical hazards, EC Category PPE 3, EC Certificate Number 0075/797/162/11/08/0462, EC Attestation Number 0075/797/162/11/08/0462
Classification Dexterity: 5
Color Yellow
Cut Resistance Rating 0
Exterior Finish Diamond
Material Latex, Cotton
Product Type Gloves
Size 9
Thickness (Metric) 0.4 to 0.8 mm
For Use With (Application) Building, Construction, Catering, Chemical, Fishing, Food, Industrial Cleaning, Laboratory, Medical, Pharmaceutical, Printing, Ship Building and Water Treatment industries
Liner Material Cotton flock
Lining Lined
Puncture Rating 0
Quantity 10Pair/PK
Tear Rating 1
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.