missing translation for 'onlineSavingsMsg'
Learn More

Fipronil Desulfinyl, TRC

Product Code. 30040960
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040960 10 mg 10mg
30040583 100 mg 100mg
30040686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040960 Supplier Toronto Research Chemicals Supplier No. TRCF34220510MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205650-65-3
Molecular Formula C12 H4 Cl2 F6 N4
Molecular Weight (g/mol) 389.08
Synonym 1H-Pyrazole-3-carbonitrile, 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)-, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)-1H-pyrazole-3-carbonitrile, Desulfinylfipronil, Fipronil desulfinyl, Fipronil metabolite 46513, MB 46513, Fipronil-desulfinyl
IUPAC Name 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)pyrazole-3-carbonitrile
SMILES Nc1c(c(nn1c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N)C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H4Cl2F6N4/c13-5-1-4(11(15,16)17)2-6(14)9(5)24-10(22)8(12(18,19)20)7(3-21)23-24/h1-2H,22H2
Formula Weight 387.9717
Chemical Name or Material Fipronil-desulfinyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.