Learn More
Justrite Fire Safety GSM
Product Code. 17702398
Change view
Click to view available options
Product Type:
Phone Dialer
Unit Size:
Each
Product Code. Product Type Unit Size
17702398 Phone Dialer Each
1 options
Product Code. 17702398 Supplier Justrite Supplier No. JCBSCEGSM0
  • ONLINE EXCLUSIVE PRICE
    856.80 €
    776.46 € / Pack of 1
    Save 80.34 €
    9% Off
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Fire Safety GSM phone dialer provides a higher level of safety in the event of a fire. The dialer continuously checks its status including signal strength, power supply, and internal operation status.
  • The Fire Safety GSM phone dialer provides a higher level of safety in the event of a fire
  • The dialer continuously checks its status including signal strength, power supply, and internal operation status and it will restart itself if there is a fault without the need for external intervention
  • The dialer outputs can be linked with alarm events and personalized profiles can be created for each number based on special requirements
  • The GSM phone dialer connects to the internet via a security procedure that can be remotely controlled allowing the dialer to be remotely programmed
  • With a 28 in. touch screen LCD display and a simple graphic interface, it makes installation and usage quick and easy
Specifications
Weight (Metric) 1 kg
Product Type Phone Dialer