missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Fireman Glove
Click to view available options
Size:
8
11
Unit Size:
Each
Description
- Heat resistant glove (formerly used as glove for firemen).
- Yellow water-repellent cowhide grain leather glove.
- Gunn cut, winged thumb, gauntlet type.
- Hand lined with Kevlar™ cotton jersey for comfort and protection from cuts and thermal hazards.
- Stitched with Kevlar™ thread for thermal insulation and sturdy seam resistance to heat and wear.
- Elasticated back.
- Velcro strap.
- Split leather cuff.
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY III Irreversible risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 407-2004 Thermal hazards, EC Category PPE 2, EC Certificate Number 0075/797/162/12/09/0517 |
| Classification | Dexterity: 5 |
| Cut Style | Gunn |
| Cut Resistance Rating | 2 |
| Material | Grain Leather, Cowhide Leather, Aramid, Cotton, Dupont™ Kevlar™ Aramid Fibers |
| Product Type | Gloves |
| Size | 11 |
| Style | Gauntlet |
| Thickness (Metric) | 1.1 to 1.3 mm |
| For Use With (Application) | Army-Defense and Fire Protection industries |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction