missing translation for 'onlineSavingsMsg'
Learn More

Fisetin, TRC

Product Code. 30404538
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404538 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404538 Supplier Toronto Research Chemicals Supplier No. TRCF35602550MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fisetin
CAS 528-48-3
Purity Grade Notes HPLC
Molecular Formula C15 H10 O6
Quantity 50 mg
Synonym 4H-1-Benzopyran-4-one, 2-(3,4-dihydroxyphenyl)-3,7-dihydroxy-, Fisetin (6CI), Flavone, 3,3',4',7-tetrahydroxy- (8CI), 2-(3,4-Dihydroxyphenyl)-3,7-dihydroxy-4H-1-benzopyran-4-one, 2-(3,4-Dihydroxyphenyl)-3,7-dihydroxy-4H-benzopyran-4-one, 3,3',4',7-Tetrahydroxyflavone, 5-Desoxyquercetin, Bois Bleu de Honqrie, C.I. 75620, C.I. Natural Brown 1, Cotinin, Fiestin, Fietin, Fisetholz, Fisitin, Fustel, Fustet, Junger Fustik, LDN 0058389, NSC 407010, NSC 656275, Superfustel, Superfustel K, Ungarisches Gelbholz, Ventin Sumach, Viset, Young Fustic, Young Fustic Crystals, Zante Fustic
InChI Formula InChI=1S/C15H10O6/c16-8-2-3-9-12(6-8)21-15(14(20)13(9)19)7-1-4-10(17)11(18)5-7/h1-6,16-18,20H
SMILES OC1=C(Oc2cc(O)ccc2C1=O)c3ccc(O)c(O)c3
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
Molecular Weight (g/mol) 286.24
Formula Weight 286.0477
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.