missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ EZ-Run™ Prestained Protein Marker
Description
- Comprised of six purified proteins covalently coupled to a blue chromophore
- Optimized to produce well-defined blue bands after SDS-PAGE
- Supplied in gel loading buffer
- 20 to 118kDa
The covalently coupled chromophore affects protein mobility; this prestained marker should only be used for approximating the molecular weight of separated proteins.
The Prestained Protein Marker is designed for monitoring protein separation during electrophoresis without staining. It is also used to verify electroblotting of proteins onto membranes.
Specifications
Specifications
| Form | Liquid |
| Formulation | Supplied in gel-loading buffer |
| For Use With (Application) | Monitoring protein separation during electrophoresis |
| Kit Contents | <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet |
| Quantity | 2 x 500 μL, 200 Loadings |
| Range | 20 to 118 kd |
| Storage Requirements | −20°C |
| Sufficient For | 200 Loadings |
| Product Type | Protein Molecular Weight Marker |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?