missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ EZ-Run™ Prestained Protein Marker

Product Code. 10714914
Change view
Click to view available options
Quantity:
2 x 500 μL, 200 Loadings
500 μL, 100 Loadings
Sufficient For:
100 Loadings
200 Loadings
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Sufficient For unitSize
10714914 500 μL, 100 Loadings 100 Loadings Each
10408923 2 x 500 μL, 200 Loadings 200 Loadings Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10714914 Supplier Fisher BioReagents™ Supplier No. BP3601500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for monitoring protein separation during electrophoresis without staining and for verifying electroblotting of proteins onto membranes

  • Comprised of six purified proteins covalently coupled to a blue chromophore
  • Optimized to produce well-defined blue bands after SDS-PAGE
  • Supplied in gel loading buffer
  • 20 to 118kDa

The covalently coupled chromophore affects protein mobility; this prestained marker should only be used for approximating the molecular weight of separated proteins.

The Prestained Protein Marker is designed for monitoring protein separation during electrophoresis without staining. It is also used to verify electroblotting of proteins onto membranes.

TRUSTED_SUSTAINABILITY

Spezifikation

Form Liquid
Formulation Supplied in gel-loading buffer
For Use With (Application) Monitoring protein separation during electrophoresis
Kit Contents <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet
Quantity 500 μL, 100 Loadings
Range 20 to 118 kd
Storage Requirements −20°C
Sufficient For 100 Loadings
Product Type Protein Molecular Weight Marker
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.