missing translation for 'onlineSavingsMsg'
Learn More

EZ-Run™ Protein Gel Solutions, Fisher BioReagents™

Resolution equal to or better than gradient precast gels

Brand:  Fisher Bioreagents BP7715-500

Code : 32

Additional Details : CAS Number : 79-06-1 Weight : 0.55000kg Transport : UN number : 3426 Chem class : 6.1 Pack group : III


 View more versions of this product

Product Code. 10284913

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

A ready-to-use premixed solution of acrylamide, bisacrylamide, buffer and SDS that enables ultra-fine resolution of protein bands by denaturing PAGE. This novel formulation provides the resolution of a gradient gel with less preparative work than a standard discontinuous gel system.

  • Ready-to-pour EZ-Run Protein Gel Solution for 1-D or 2-D PAGE—just add APS and TEMED
  • Wide separation range—from 3 to 200kDa on same minigel
  • Continuous gel system—no stacking gel necessary
  • Proprietary (patent-pending) gel chemistry

EZ-Run Protein Gel Solution comes with an optimized proprietary running buffer; other traditional SDS-PAGE buffers should not be used with the EZ-Run gel solution.

Applications:

Compatible with post-electrophoresis applications, such as Western blotting, MALDI analysis, protein sequencing. Suitable for all commonly used gel staining methods including Coomassie™ blue, silver staining and fluorescent dyes.

Specifications

Specifications

Protein Gel Solution
15% protein gel solution
15%
500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 15% EZ-Run Protein Gel Solution with buffer

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H372-Causes damage to organs.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361f-Suspected of damaging fertility.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents

Documents

Certificates
Special Offers

Special Offers