missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ EZ-Run™ Protein Gel Staining Solution

Product Code. 10786444
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10786444 1 L 1 L
10609933 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10786444 Supplier Fisher BioReagents™ Supplier No. BP36201
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ready-to-use solution for highly sensitive staining of proteins separated in polyacrylamide gels

EZ-Run Protein Gel Staining Solution contains Coomassie™ Brilliant Blue G-250. Due to the property of the dye to form colloidal particles, proteins are preferentially stained without significant background staining of the gel matrix.

  • Detection is linear over two orders of magnitude with as little as 5ng protein detected
  • Sensitivity: 5ng BSA in 10% SDS-PAGE, 1.0mm thick gel, well width 3mm
  • Does not contain toxic methanol or acetic acid
  • Provides rapid destaining with simple water washes
  • Can be reused twice without significant loss of sensitivity
  • Tested for staining sensitivity in minigels using purified proteins and Protein Molecular Weight Standards

TRUSTED_SUSTAINABILITY

Specifications

Color Dark Blue
Form Liquid
Formulation Coomassie™ Brilliant Blue G-250
For Use With (Application) Highly sensitive staining of proteins separated in Polyacrylamide gels
Quantity 1 L
Product Type Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.