missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ EZ-Run™ Protein Marker
Description
- Supplied in gel loading buffer for direct application to SDS-PAGE gels
- Ideal for molecular weight determination of polypeptides analyzed in various gel systems
Specifications
Specifications
| Form | Liquid |
| Formulation | Supplied in gel-loading buffer |
| For Use With (Application) | Monitoring protein separation during SDS-PAGE |
| Kit Contents | <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet |
| Quantity | 500 μL, 100 Loadings |
| Range | 14.4 to 116 kd |
| Storage Requirements | −20°C |
| Sufficient For | 100 Loadings |
| Product Type | Protein Molecular Weight Marker |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?