missing translation for 'onlineSavingsMsg'
Learn More

Granulated Agar, Fisher BioReagents

Product Code. 15470665
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging Type:
Poly Bottle
Poly Pail
Unit Size:
2kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15470665

Brand: Fisher BioReagents BP97442

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used as a solidifying agent to provide the solid medium to support the growth of microbes

  • Granulated agar media CAS:[9002-18-0]
  • Store at room temperature

TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Product Type Agar
For Use With (Application) Used as a solidifying agent to provide the solid medium to support the growth of microbes
Packaging Type Poly Pail
Quantity 2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.