missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ exACTGene™ Control Primers
Optimized for routine PCR applications
4.40 € - 4.45 €
Specifications
Content And Storage | -20°C |
---|---|
For Use With (Equipment) | Fisher BioReagents™ exACTGene™ PCR kit |
Quantity | 1 Kit |
Concentration | 20μm |
Volume | 100μL |
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
10707034
|
Fisher BioReagents™
BP6118 |
Control Primer No. 3 | 1 Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10061553
|
Fisher BioReagents™
BP6116 |
Control Primer No. 1 | 1 Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11428293
|
Fisher BioReagents™
BP6117 |
Control Primer No. 2 | 1 Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
-20°C | |
1 Kit | |
100μL |
Fisher BioReagents™ exACTGene™ PCR kit | |
20μm |