missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ exACTGene™ DNA Ladders

Product Code. 10657633
Change view
Click to view available options
For Use With (Application):
Clone Identification
Fast run times, small size DNA
General purpose, extra large size
General purpose, large digested DNA
General purpose, small size DNA
General purpose, wide size range
Large size PCR or cloning
PCR size confirmation
Quick check of PCR or Enzyme digestion
Small and large cloning
No. of Bands:
8
10
11
12
13
14
15
16
19
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) No. of Bands unitSize
10657633 PCR size confirmation 12 Each
10224973 Quick check of PCR or Enzyme digestion 8 Each
10061413 General purpose, small size DNA 10 Each
10214973 PCR size confirmation 14 Each
10021463 Fast run times, small size DNA 11 Each
10306943 Clone Identification 14 Each
10031463 Large size PCR or cloning 10 Each
10122823 Small and large cloning 16 Each
10489883 General purpose, large digested DNA 13 Each
10699163 General purpose, extra large size 15 Each
10499883 General purpose, wide size range 19 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10657633 Supplier Fisher BioReagents™ Supplier No. BP2571100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze

  • Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
  • Ideal for size assessment and quantitative determination of DNA mass
  • Contain loading dye to reduce pipetting steps and save time
  • Have two or three high intensity reference bands for better sizing
  • Provided in sufficient quantity to load 100 lanes

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Form Liquid
Formulation Premixed with loading dye
For Use With (Application) PCR size confirmation
No. of Bands 12
Quantity 100 Lanes
Storage Requirements Stable at room temperature for two years.
Sufficient For 100 Lanes
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.