missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ exACTGene™ Lambda DNA Template

Product Code. 11408293
Change view
Click to view available options
:
Lambda DNA Control Template, 100μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11408293 Lambda DNA Control Template, 100μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408293 Supplier Fisher BioReagents™ Supplier No. BP6115
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Optimized for routine PCR applications

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 ng/μL
Content And Storage -20°C
For Use With (Equipment) Fisher BioReagents™ exACTGene™ PCR kit, troubleshoot PCR parameters
Product Type Lambda DNA Template (1ng/μL)
Volume 100μL
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.