missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ LB Agar, Lennox (Granulated)
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Poly Bottle
Poly Pail
Unit Size:
2kg
500g
Description
- Granulated LB Agar, Lennox media
- CAS: [91079-40-2], [8013-01-2], [7647-14-5], [9002-18-0]
- Store at room temperature
Specifications
Specifications
| Form | Granulated Powder |
| Product Type | Agar |
| For Use With (Application) | Used as a solidifying agent to provide the solid medium to support the growth of microbes |
| Packaging Type | Poly Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction