missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Agar, Miller (Granulated)

Product Code. 12887172
Change view
Click to view available options
Quantity:
2 kg
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12887172 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12887172 Supplier Fisher BioReagents™ Supplier No. BP97242
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 32g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
  • Agar provides solid support for growth
  • Format: Granulated powder
TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Media Type LB Agar, Miller
Product Type Nutrient Agar
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 12g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Bottle
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.