missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Leupeptin Hemisulfate

Product Code. p-3138483
Click to view available options
Quantity:
1 mg
5 mg
25 mg
100 mg
Unit Size:
100mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 10757024

Brand: Fisher BioReagents™ BP26621

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

(Acetyl-Leu-Leu-Arg-al), Valeramide

A biochemical used as an inhibitor for trypsin, plasmin, papain, and cathepsin B.

TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Inhibitor for Trypsin, Plasmin, Papain, Cathepsin B
Content And Storage -20°C
Quantity 1 mg
Product Type Leupeptin Hemisulfate
Molecular Weight (g/mol) 475.60
Form Chunks, Powder
compliance-icons
Product Identifier
  • Leupeptin Hemisulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.