missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Microbiology Media Additives: Yeast Extract

Product Code. 10215203
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 10215203

Brand: Fisher BioReagents™ BP14222

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Additive for preparing bacterial nutrient media

Water soluble portion of autolyzed fresh yeast used in bacteriological culture media
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Grade Molecular genetics
Media Type Yeast Extract
Product Type Microbiology Media
Packaging Type Poly Bottle
Quantity 2000 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.