missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Microbiology Media Additives: Yeast Extract

Product Code. 10255153
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10255153 100 g 100g
10225203 500 g 500g
10215203 2000 g 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255153 Supplier Fisher BioReagents™ Supplier No. BP1422100
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Additive for preparing bacterial nutrient media

Water soluble portion of autolyzed fresh yeast used in bacteriological culture media
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Grade Molecular genetics
Media Type Yeast Extract
Product Type Microbiology Media
Packaging Type Poly Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.