missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Molecular Weight Markers

Product Code. 11483084
Change view
Click to view available options
Quantity:
10 μg
25 μg
50 μg
Product Type:
100 bp DNA Ladder
Kilobase DNA Marker
PhiX-174 RF DNAHae III Digest
lambda DNA/Hind III/PhiX-174 RF DNA-Hae III Digest
Unit Size:
1µg
50µg
Size Range:
100 to 2000 bp
500 to 10000 bp
72 to 1353 bp
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize Size Range
11483084 25 μg lambda DNA/Hind III/PhiX-174 RF DNA-Hae III Digest 1µg -
10428543 50 μg 100 bp DNA Ladder 50µg 100 to 2000 bp
10523325 50 μg Kilobase DNA Marker 1µg 500 to 10000 bp
11468473 10 μg PhiX-174 RF DNAHae III Digest 1µg 72 to 1353 bp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11483084 Supplier Fisher BioReagents™ Supplier No. BP255425
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for determining the size of DNA fragments

  • Compatible with both conventional and pulsed field gel electrophoresis
  • DNase-, RNase-, and protease-free

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Form Lyophilized
For Use With (Application) Determining the size of DNA fragment
Quantity 25 μg
Product Type lambda DNA/Hind III/PhiX-174 RF DNA-Hae III Digest
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.