missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Proteinase K (Tritirachium album/Molecular Biology)

Product Code. 10103533
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10103533 100 mg 100mg
10172903 50 mg 50mg
10407583 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103533 Supplier Fisher BioReagents™ Supplier No. BP1700100
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy

Proteinase K (Tritirachium album/Molecular Biology)

  • Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes

Proteinase K is widely used in the isolation of DNA and RNA from cell and tissue preparations. It is a wide-spectrum protease, inactivating many enzymes, including endogenous nucleases.

TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases
Content And Storage -20°C
Source Tritirachium album
Quantity 100 mg
Product Type Proteinase K
Form Solid
compliance-icons compliance-icons
Product Identifier
  • Proteinase K
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.