missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Restriction Enzymes - Bgl I (Source: Bacillus globigii)
Description
- Application: Bgl I restriction endonuclease digests dsDNA with sequence indicated below
- Storage Buffer Components: 10mM Tris-HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA and 50% glycerol
- Storage: -20°C
Specifications
Specifications
| Concentration | 10 U/μL |
| Quantity | 1000 units |
| Cut Site | GCCNNNN.NGGC |
| Product Type | BgII |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?