missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Restriction Enzymes - Xba I (Source: Xanthomonas badrii)

Product Code. 10173483
Change view
Click to view available options
Quantity:
2000 U
Unit Size:
2000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10173483 2000 U 2000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10173483 Supplier Fisher BioReagents™ Supplier No. BP34481
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

•    Application: Xba I restriction endonuclease digests dsDNA with sequence indicated
•    Storage Buffer Components: 10mM Tris-HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA and 50% glycerol
•    Qualified for Blue/White Cloning Assays and genomic assays
•    Storage: -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 8 to 12 U/μL
Quantity 2000 U
Cut Site T.CTAGA
Product Type XbaI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.