Learn More
Fisher BioReagents™ SurePrep™ Urine Exfoliated Cell RNA Purification Kit
Description
Includes
Lysis solution (40mL); Wash solution (22mL); RNA elution solution (6mL); Spin columns (50); Collection tubes (50); Elution tubes (1.7mL) (50); Product insert (1)
- Fast and easy processing using rapid spin-column format
- Isolate total RNA, from large mRNA down to microRNA (miRNA)
- RNA can be isolated and detected from as little as 100 exfoliated cells
- Isolate high quality total RNA from urine
- No phenol or chloroform extractions
Applications:
Purified RNA from urine exfoliated cells suitable for real-time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays
Specifications
Specifications
| Format | Spin Column |
| For Use With (Application) | Purified RNA from urine exfoliated cells suitable for RT-PCR, reverse transcription PCR, northern blotting, RNase protection and primer extension and expression array assays |
| Includes | Lysis solution 40mL, wash solution 22mL, RNA elution solution 6mL, spin columns 50, collection tubes 50, 50 elution tubes 1.7mL, product insert 1 |
| Quantity | 50 Preps |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.