missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ 12mm Karl Fisher Electrode Bulb Protector
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Accumet 12 mm dia. Electrode
Unit Size:
Each
Description
Protect fragile glass membrane against breakage
Protectors slip over electrode tip to guard glass bulb
Specifications
Specifications
| For Use With (Equipment) | Accumet 12 mm dia. Electrode |
| Type | Bulb Protector |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction