missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AP115 Portable pH Meter Kit

Product Code. 11570184 Shop All Fisherbrand Products
Change view
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
Product Code. Range (pH) unitSize
11570184 -2 to 20 Each
1 options
This item is not returnable. View return policy

Product Code. 11570184

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Meter, accumet combination refillable pH/ATC electrode 13-620-AP50A, refill solution, hard carrying case, sample bottles, pH calibration packets, 9V battery and manual.

Accurate measurements any time, anywhere

For indoor and outside use. Versatile and rugged enough for the most demanding applications. Soft touch keys with comfort grip make one-handed operation a breeze. Compact design is small enough for a shirt pocket yet smart enough for any laboratory. Kit includes an unbreakable accumet pH/ATC electrode that is refillable for fast response and longest life.

AP115 pH/mV/Temp meter offers all features of the AP110 meter, plus:

  • 0.001pH resolution
  • Date/time display to meet GLP requirements

TRUSTED_SUSTAINABILITY

Specifications

Includes Meter, Accumet™ combination refillable pH/ATC electrode 13-620-AP50A, refill solution, hard carrying case, sample bottles, pH calibration packets, 9V battery and manual
For Use With (Application) pH, mV, Temperature
Range (pH) -2 to 20
Resolution (pH) 0.001
Range (mV) 0 to ±2000
Temperature Range (Metric) -5°C to +100°C
Display Type LCD
Certifications/Compliance GLP
Warranty 3 Years
Accuracy Multiple
Resolution 0.001pH
Detection Range -2 to 20 pH
Type AP115 Portable pH Meter Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.