missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AP85 Portable Waterproof pH/Conductivity Meter

Product Code. 11500254 Shop All Fisherbrand Products
Change view
Click to view available options
Calibration Points:
5
Unit Size:
Each
Product Code. Calibration Points unitSize
11500254 5 Each
1 options
This item is not returnable. View return policy

Product Code. 11500254

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Meter/Probe package includes meter, 1.0cm-1 conductivity/temperature plastic-bodied probe with 3 ft. (0.9m) long cable (13-620-AP54), All-in-One pH/temperature probe (13-620-AP55), four AAA batteries and instruction manual.

Meter Kit package includes meter, 1.0cm-1 conductivity/temperature plastic-bodied probe with 3 ft. (0.9m) long cable (13-620-AP54), All-in-One pH/temperature probe (13-620-AP55), 4.00 and 7.00pH buffer solutions, 1413μS conductivity calibration solution, sample bottle, four AAA batteries, instruction manual and hard plastic carrying case.

Durable, waterproof, dustproof—ideal for rough field applications

Measures pH, conductivity and TDS plus temperature. Switches between pH, conductivity and TDS with the press of a button. Dual-display LCD shows all parameters with temperature; recalls previous calibration points too.

  • Advanced set-up mode for customizing meter parameters
  • Selectable automatic endpoint freezes reading immediately after stabilization
  • HOLD button freezes reading
  • Self-diagnostic: meter indicates when it is ready to operate; error messages alert operator to problems
  • ISO™ 9001 certified manufacturer

Powerful pH Mode
  • Recalls pH electrode slope and offset
  • Has five-point calibration with automatic buffer recognition

Conductivity and TDS Modes
  • Recalls and customizes effective cell constants for each range
  • Autoranging for conductivity across five ranges
  • Autoranging for TDS across six ranges
  • Five-point calibration for ±1% full-scale accuracy

Flexible Temperature Mode
  • Select temperature in °C or °F Set coefficient from 0 to 10% for accurate compensation in almost any solution
  • Manual or automatic temperature compensation

Service Plan

Service agreement available.

Warranty

accumet Three-Year Warranty. accumet AP85 is warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years. Call your Fisher Scientific Customer Service Representative for full details.

TRUSTED_SUSTAINABILITY

Specifications

Includes Meter, 1.0cm-1 conductivity/temperature Plastic-bodied probe with 3 ft. (0.9m) long cable (13-620-AP54), All-in-one pH/temperature probe (13-620-AP55), 4.00 and 7.00pH buffer solutions, 1413μS conductivity calibration solution, sample bottle, four AAA batteries, carrying case
Accuracy (Conductivity) ±1% Full-scale
For Use With (Application) pH, Conductivity, TDS
Cell Constant 1cm-1
Calibration Points 5
Range (Conductivity) 0 to 19.99 μS, 0 to 199.9 μS, 0 to 1999 μS, 0 to 19.99 mS, 0 to 199.9 mS
Resolution (Conductivity) 0.01μS, 0.1μS, 1μS, 0.01mS, 0.1mS
Temperature Compensation Automatic or Manual
Range (TDS) 0 to 9.99 ppm, 10 to 99.9 ppm, 100 to 999 ppm, 1 to 9.99 ppt, 10 to 99.9 ppt, 100 to 200 ppt
Resolution (TDS) 0.01 ppm, 0.1 ppm, 1 ppm, 0.01 ppt, 0.1 ppt, 1 ppt
Accuracy (TDS) ±1% Full-scale
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Accuracy (Temperature) ±0.5°C
Display Type LCD
Certifications/Compliance ISO 9001
Weight (English) 0.04 oz.
Warranty 3 Years
Resolution 0.01 pH
Model Meter Kit Package
Sensor Types Conductivity
Detection Range -2 to +16pH
Accuracy +0.01 pH
Length (Metric) 19 cm
Width (English) 3.75 in.
Width (Metric) 9.5 cm
Height (English) 2.25 in.
Battery Type 4 x AAA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.