missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Automatic Temperature Compensation Probes - Mercury - Free

Product Code. 10236064 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Connector Types:
0.25 in. Phone
Mini-Phone
Material:
316 Stainless Steel
Stainless Steel
For Use With (Equipment):
Accumet benchtop and portable meter
Accumet model AP63, AP62, AP61
accumet AB, AR, XL, Basic, benchtop meters
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Material For Use With (Equipment) Unit Size
10236064 Mini-Phone 316 Stainless Steel accumet AB, AR, XL, Basic, benchtop meters Each
11530234 0.25 in. Phone Stainless Steel Accumet benchtop and portable meter Each
11540234 Mini-Phone Stainless Steel Accumet model AP63, AP62, AP61 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10236064
  • ONLINE EXCLUSIVE PRICE
    266.00 €
    167.65 € / Pack of 1
    Save 98.35 €
    36% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For accumet™ benchtop meters.

For use with accumet Meters

  • Signal solution temperature to meter for automatic compensation
  • Permit temperature readout with meters having this feature
  • Probes for accumet AP Series portable meters have waterproof connector
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) accumet AB, AR, XL, Basic, benchtop meters
Material 316 Stainless Steel
Temperature (Metric) Operating 0°C to 100°C
Type ATC
Connector Types Mini-Phone
Length (Metric) Cable 0.9 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.