missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Polymer Membrane Combination ISEs, Mercury-Free: Potassium

Product Code. 11510144 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11510144 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11510144
  • ONLINE EXCLUSIVE PRICE
    847.00 €
    630.16 € / Pack of 1
    Save 216.84 €
    25% Off
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fast, precise measurements of calcium, nitrate and potassium ion in aqueous solutions over a wide range of concentrations

Fast, precise measurements in aqueous solutions over a wide range of concentrations.

Sensor sealed in epoxy body

  • Sensing modules contain liquid internal filling solution in contact with gelled organophilic membrane
  • Membrane is impregnated with a selective ion exchanger for ion of interest
  • Sensors are sealed into durable epoxy body with built-in reference half-cell
  • Permit fast, precise measurements in aqueous solutions over a wide range of concentrations
  • Epoxy body provides chemical resistance
Features
  • Tough, chemical-resistant epoxy body
  • Membrane assembly sealed to body
  • Continuous electrical shielding and insulation of internal elements, cable and connectors
  • Extremely stable, reproducible readings with a minimum of electrical noise
  • Lower detection limits than conventional ISEs
  • Rapid response time that lowers cost per test
  • Individually tested, serialized to meet GLP requirements and backed by the Fisher Scientific Technical Applications staff
Dimensions
  • Diameter: body, 0.5 in. (12.7mm); cap, 0.625 in. (16mm)
  • Length: body, 4 in. (10.2cm); cap, 1 in. (2.5cm); overall, 5 in. (12.7cm) 

Potassium Electrode

  • Aids in analysis of serum, urine, saliva, other bodily fluids
  • Used in testing soils, sewage, fertilizers, foods and beverages
  • Filling solution is 5M NaCl

 

TRUSTED_SUSTAINABILITY

Specifications

Applications Used in testing soils, sewage, fertilizers, foods, beverages, analysis of serum, urine, saliva and other bodily fluids
Connector Types BNC
Diameter (English) Body 0.5 in.
Diameter (Metric) Body 12.7mm
Diameter (Metric) Cap 15.8mm
Filling Solution Type 4mol/L KCl with AgCl
Length (English) 5 in.
Length (Metric) 127mm
Material Epoxy
Operating Range (Metric) 0.04 to 39000ppm
Temperature (English) Operating 32° to 88°F
pH Range 2.5 to 11
Interferences Cs+, NH4, Tl+, H+, Ag+, Tris+, Li+, Na+
Resistance 10 to 20megohm
Response Time ∽1 min. or less
Electrode Potassium
Temperature (Metric) Range 0 to 40°C
Temperature (Metric) Operating 0°C to 40°C
Type Membrane Combination ISE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.