Learn More
Fisherbrand™ DAS-3 Data Acquisition System for Traceable™ Products with Computer Output
Product Code. 11704286 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Digital thermometer, humidity/temperature/dew point meter, Platinum ultra accurate thermometer Stainless Steel probe
Manometer/pressure/vacuum gauges, memory hygrometer/thermometer, expanded range conductivity meter and memory hygrometer/thermometer
Memory wide range thermometer, general purpose hygrometer/thermometer and anemometer/thermometer
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11704286 Digital thermometer, humidity/temperature/dew point meter, Platinum ultra accurate thermometer Stainless Steel probe Each
11734316 Memory wide range thermometer, general purpose hygrometer/thermometer and anemometer/thermometer Each
11764316 Manometer/pressure/vacuum gauges, memory hygrometer/thermometer, expanded range conductivity meter and memory hygrometer/thermometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11704286
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
Includes
CD, 6 ft. cable (supplied USB, serial, and instrument connections) cable plugs into the instrument and computer.
Powerful and easy to use computer data capture program works with Traceable Instruments with computer output—designed to work with Windows™ 98/Me/NT/2000/XP/Vista/Windows 7. Connects up to eight instruments all running simultaneously on one computer.
  • Capture readings permanently
  • Records in intervals from 1 to 10,000 seconds
  • Straightforward plug-and-play
  • Displays minimum/maximum readings
  • Utilizes an alarm mode that permits the user to be notified visually, audibly, and by email when an alarm is triggered
  • Unlimited readings are saved to a file that can be viewed and printed as is, or may be printed in any report or spreadsheet format
  • Networking server/client capability allows the captured data to be monitored on a remote workstation
  • Readings may be streamed to cell phones or anywhere via email every minute or in any time interval
  • 24/7 notification of audible/visual alarms are sent to user's computer, remote computer, via email anywhere, and by email to cell phones
  • Program automatically installs in less than two minutes—no user programming, no user keystrokes, no user entries required: simply connect the cable to the instrument and to a USB port (or serial port) and data is captured
Specifications
Type Computer Data Acquisition System
Includes CD, 6 ft. USB cable
For Use With (Application) Capture readings permanently, displays minimum/maximum readings
For Use With (Equipment) Digital thermometer, humidity/temperature/dew point meter, Platinum ultra accurate thermometer Stainless Steel probe