missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hands-Free Fold-Up Magnifiers

Product Code. 11744396 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification:
10x
2.5x
3.5x
3x
5x
7x
Diameter (Englis) Lens:
0.5 in.
0.87 in.
1.25 in.
2 in.
3 in.
3.5 in.
4 in.
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Diameter (Englis) Lens unitSize
11744396 2.5x 4 in. Each
11754396 2.5x 3.5 in. Each
11764396 3x 3 in. Each
11704406 10x 0.5 in. Each
11784396 5x 1.25 in. Each
11774396 3.5x 2 in. Each
11794396 7x 0.87 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11744396

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Batteries, with lighted models.

Allow two-handed examination of objects while yielding a sharp, crisp and distinct image

Fisher Scientific™ Hands-Free Fold-Up Magnifiers all fold twice for compact storage. Allow two-handed examination of objects while yielding a sharp, crisp and distinct image.

  • Bottom panel features white grid markings in both inches and millimeters to assist in measuring
  • All-glass optics ensure perfect focus with zero distortion
  • Seven sizes are available, including three featuring brilliant battery-powered illumination

Specifications

For Use With (Application) For use in quality control, labs, food, electronics, manufacturing and universities
Includes Battery
Diameter (Englis) Lens 4 in.
Diameter (Metric) Lens 10.16 cm
Magnification 2.5x
Material Glass
Product Type Magnifier (Hands-free, Fold-up)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.