missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters

Product Code. 15387928 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
No. of Places:
1
2
4
Plug Type:
CN, EU, UK
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Places Plug Type Unit Size
15387928 1 CN, EU, UK Each
15397928 2 CN, EU, UK Each
15317938 4 CN, EU, UK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15387928
  • ONLINE EXCLUSIVE PRICE
    462.00 €
    417.00 € / Pack of 1
    Save 45.00 €
    9% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

Requires

Unit must be ordered with block(s)

Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.

  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment

Applications:

Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation

Warranty

2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, cULus, RoHS
Controller Type PID
Material Powder-coated Steel
No. of Places 1
Plug Type CN, EU, UK
Relative Humidity <80%
Temperature Range (Metric) Ambient +5° to 130°C
Type Dry Bath Block
Fuse 250V 2.5A
Heat Up Time ≤ 20 min. 30° to 130°C
Includes Dry bath, CN, EU and UK plugs, block lifter
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 1°C
Timer 0 to 99:59 min or continuous
Warranty 2 Year, Replacement
Dimensions (L x W x H) 11.3 x 7.9 x 3.9 in. (288 x 200 x 100 mm)
Length (English) 11.3 in.
Length (Metric) 288 mm
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 3.9 in.
Height (Metric) 100 mm
Weight (English) 6.39 lb.
Weight (Metric) 2.9 kg
Electrical Requirements 200/240 V, 50/60 Hz (CN, EU, UK)
Voltage 200/240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.