missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cover for Precision™ Water Baths, 4 holes, for 10L Water Bath

Product Code. 15355897 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15355897 10 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15355897 Supplier Fisherbrand™ Supplier No. 1546230Q
  • ONLINE EXCLUSIVE PRICE
    768.83 €
    692.00 € / Pack of 1
    Save 76.83 €
    9% Off
Estimated Shipment: 22-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These Fisherbrand™ Covers are for use with Precision Dubnoff and Shallow-Form Shaking Water Baths.

Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
TRUSTED_SUSTAINABILITY

Specifications

Description Concentric Ring Cover 4 Holes
Capacity (Metric) 10 L
For Use With (Equipment) Precision 10L General Purpose Water Baths
Material Stainless Steel
No. of Positions 4
Warranty 1 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.