missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Conductivity, Resistivity, and TDS Meter

Product Code. 11774216 Shop All Fisherbrand Products
Change view
Click to view available options
Display Type:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type unitSize
11774216 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11774216

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Provides direct digital readings of conductivity, resistivity and dissolved solids. Traceable™ Conductivity, Resistivity, and TDS Meter is ideal for measuring reagent-grade water to meet CAP, ASTM, NCCLS, and ACS specifications as well as for measuring conductivity in chromatography, biology, food, electronics, and water analysis. All readings automatically referenced to the international standard of 25°C.

  • Bright, easy-to-read, 0.5 in. (13 mm) LCD
  • Can be used as handheld or benchtop meter
  • Ranges: 0.01 to 200,000 μmhos or μS (conductivity); 0.001 ohms to 20.00 megohms (resistivity); 0.01 to 20,000 ppm (dissolved solids)
  • Accuracy, +0.3% ± 1 digit
  • Stores four calibration settings
  • With plug-in, instant-response probe, digital temperature display in °C and °F, automatic temperature compensation and control, and recorder jack

Certifications

Individually serial-numbered Traceable™ Certificate from an ISO 17025 calibration laboratory accredited by A2LA indicating instrument traceability to standards provided by NIST™.

Specifications

Accuracy (Conductivity) ±0.3% ±1 digit
For Use With (Application) Conductivity, Resistivity, TDS
Measurement Modes Conductivity, TDS, Resistivity
Range (Conductivity) 0.1 to 200 μS
Range (Resistivity) 1 to 20 megohm
Display Type LCD
Certifications/Compliance ISO 17025, NIST
Weight (Metric) 454 g
Weight (English) 0.03 oz.
Sensor Types Conductivity
Detection Range 0.01 to 200,000 μS
Type Conductivity Meter
Accuracy ±0.3%
Length (English) 3.75 in.
Length (Metric) 9.5 cm
Length (Metric) Cable 150 cm
Width (English) 16 oz.
Width (Metric) 17.1 cm
Height (English) 1.5 in.
Height (Metric) 3.8 cm
Battery Type Alkaline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.