missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Dual-Display Digital Stopwatch

Product Code. 11769825 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
Stopwatch; lanyard; Battery
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
11769825 Stopwatch; lanyard; Battery Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11769825

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Stopwatch; lanyard

Engineered for ease of operation, accuracy, and reliability. Fisherbrand Traceable Dual-Display Digital Stopwatches provide precise, Traceable timing for enzyme tests, viscosity measurements, animal studies, and all timed lab tests. They are made for scientists in industrial, educational, hospital, and governmental labs.

Engineered for ease of operation, accuracy, and reliability. Fisher Scientific™ Traceable™ Dual-Display Digital Stopwatches provide precise, Traceable timing for enzyme tests, viscosity measurements, animal studies, and all timed lab tests. They are made for scientists in industrial, educational, hospital, and governmental labs. Quartz-crystal design is accurate to 0.0005%—over 100 times more accurate than other stopwatches.

Set countdown time from 10 hours to 1 second. When zero is reached, timer alarms and repeats countdown from the programmed time. Repeat cycle automatically continues until stopped. Display count box shows how many times the cycle has been repeated. LCD screen continually displays time remaining to zero. 

  • Functions include single-action, time-out, interval-split, cumulative split, continuous roll-over timing, and countdown alarm timing
  • Dual display shows cumulative and interval split
  • Features eight recallable memories, plus fastest and slowest lap times
  • Repeat timer provides repeat function of countdown alarm timing
  • Tough, water-resistant ABS plastic case seals out moisture, dust, and fumes encountered in harsh lab, plant, or field environments
  • Timing range is 9 hours, 59 minutes, 59 seconds, and 99 hundredths
  • Resolution is 0.01 second
  • Accuracy is 0.0005%
  • Size: 6.4 x 7.6 x 1.9 cm (2.5 x 3 x 0.75 in.)
  • Weight: 2.5 oz.

Order Info

Replacement battery: Cat. No. 02-440-1A

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.

Specifications

Product Type Dual-Display Digital Stopwatch
Time Range 10 hr.
Resolution 0.01 sec.
Display LCD
Dimensions (L x W x H) 2.5 x 3 x 0.75 in. (6.35 x 7.62 x 1.9 cm)
Length (English) 2.5 in.
Length (Metric) 6.35 cm
Width (English) 2 in.
Width (Metric) 5.08 cm
Height (English) 0.75 in.
Height (English) Display 0.25 in.
Height (Metric) 1.9 cm
Weight (English) 2.5 oz.
Weight (Metric) 71 g
Includes Stopwatch; lanyard; Battery
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.