Learn More
Fisherbrand™ Traceable™ Pure H2O Tester
Product Code. 11794216 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Range:
0.1 to 20000μS
Unit Size:
Each
Product Code. Range Unit Size
11794216 0.1 to 20000μS Each
1 options
Product Code. 11794216
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Includes
Probe with 40 in. cable; a Traceable Certificate to indicate instrument traceability to NIST from an ISO 17025 calibration laboratory; carrying case; instructions and 9V battery.
Single-purpose unit designed for labs required to maintain a periodic check of water purity
Accurate readings in an instant. Use to test water from stills, demineralizers, deionizers, and reverse osmosis equipment.
  • Simple operation eliminates errors
  • Internal, solid-state thermometer automatically references readings to international standard of 25°C (77°F)
  • Accuracy is ±0.4% of full scale
  • Range is 0.1 to 20,000 micromhos (from pure to raw water)
  • Comes ready to operate
Compliance
Meets all lab certification requirements for pure water analysis for CAP, ASTM™, NCCLS, USP, and ACS.
Specifications
Certifications/Compliance CAP, ASTM, NCCLS, USP, and ACS
For Use With (Application) Single purpose unit designed for labs required to maintain a periodic check of water purity
For Use With (Equipment) Demineralizers, deionizers, reverse osmosis equipment
Includes Probe with 40 in. cable, traceable™ certificate to indicate instrument traceability to NIST from an ISO 17025 calibration laboratory, carrying case, instructions and 9V battery
Range 0.1 to 20000μS
Type Traceable™ Pure H2O Tester