missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ RTD Platinum Thermometer

Product Code. 11729765 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Temperature Range (Metric):
-100°C to +300°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
11729765 -100°C to +300°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11729765
  • ONLINE EXCLUSIVE PRICE
    436.00 €
    396.33 € / Pack of 1
    Save 39.67 €
    9% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Includes

9V battery and probe with 40 in. (102cm) cable

High-precision, durable thermometer with platinum RTD probe, and linear fuzzy logic chip for wide temperature-range operation

Platinum (100ohm) four-wire RTD probe can be used in liquids, air/gas, and semisolids.

  • Resolution: 0.01° below 200° and 0.1° above 200°
  • Accuracy: ±(0.1% + 0.2°C) below 200°, ±(0.15% + 0.5°C) above 200°
  • Probe range: -100° to +300°C
  • Features HOLD button to freeze display and capture reading
  • Fail-safe display indicates low-battery and probe condition
  • Easy-to-read, high-contrast LCD with 0.51 in.H (1.3cm) digits
  • Readings are updated 2.5 times every second
  • Ideal for experimental applications and in severe environments
  • Reproducible one-button operation minimizes operator error in quality control environments
  • Use to measure temperatures in soil, foods, ovens, cuvets, gas systems, reagents, water baths, wastewater, incubators, chemical solutions, petroleum products, and extreme cold

Order Info

Unit dimensions: 1.25L x 3W x 7 in.H (3.2 x 7.6 x 17.8cm); Weight: 9.5 oz. (269g); Probe measures 6 in.; Overall length: 12 in.

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Specifications

Calibration Points 0, 50, and 100°C
Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.13 in.
Dimensions (L x W x H) 4 x 3 x 7 in. (10.1 x 7.6 x 17.8 cm)
For Use With (Application) Lab and plant use
Height (English) 7 in.
Height (Metric) 17.8 cm
Includes 9 V Battery and probe with 40 in. (102cm) cable
Length (English) Probe 9 in.
Length (Metric) 101 mm
Length (Metric) Probe 22.8 cm
Material Plastic
Resolution 0.1°C
Weight (English) 9.5 oz.
Weight (Metric) 200 g
Width (English) 3 in.
Product Type Digital Thermometer
Length (English) 4 in.
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -148°F
Min. Temperature (Metric) -100°C
Temperature Range (Metric) -100°C to +300°C
Accuracy 0.5°C
Display Digital LCD
Width (Metric) 7.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.