Learn More
Fisherbrand™ Vortex Mixer Accessories: Ampule Tube Holders
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15716959
  • 120.00 € / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Mixes storage vials and test tubes
Specifications
Specifications
15 to 17 mm dia.
Ampoule Holder
Vortex mixer