missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Horizontal Minigel Systems: Combs

Product Code. 11902955
Change view
Click to view available options
Thickness (Metric) Teeth:
1 mm
1.5 mm
Volume (Fill) Metric:
55 μL
80 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Teeth Volume (Fill) Metric unitSize
11902955 1.5 mm 80 μL Each
12434567 1 mm 55 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11902955 Supplier Fisherbrand™ Supplier No. B1A5D
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Horizontal Minigel systems

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Owl™ EasyCast™ B1A mini Gel and A2-OK multi Gel Electrophoresis System
Type Standard Comb
Teeth Count 5
Thickness (Metric) Teeth 1.5 mm
Volume (Fill) Metric 80 μL
Width (Metric) Teeth 11.3 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.