Learn More
Fisherbrand™ 200 Series Safety Glasses
Economical, lightweight protection
89.01 €
Specifications
| Certifications/Compliance | ANSI Z87.1, ANSI Z80.3 |
|---|---|
| Lens Tint | Clear |
| Product Type | Safety Glass |
| Temple Style | Adjustable; spatula |
Description
Economical, lightweight protection
- One-piece lens provides a full and unobstructed field of vision
- Absorbs >99.9% of harmful UV light
- Molded-in sideshields and browguards
- Adjustable spatula temples
Certifications
CSA approved.
Compliance
Meets ANSI* Z87.1 and Z80.3 standards.
Specifications
| ANSI Z87.1, ANSI Z80.3 | |
| Clear | |
| Safety Glass | |
| Adjustable; spatula |